Anti-KRTAP13-1 Mouse Monoclonal Antibody [clone: 2A1]

Supplier: US Biological
USBI128970EA 702 CHF
USBI128970
Anti-KRTAP13-1 Mouse Monoclonal Antibody [clone: 2A1]
Antibodies
Type: Primary
Antigen: KRTAP13-1 (keratin associated protein 13-1)
Clonality: monoclonal
Clone: 2A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR