Anti-KRT83 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI128968EA 797 CHF
USBI128968
Anti-KRT83 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KRT83 (keratin 83)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR