Anti-KRT81 Mouse Monoclonal Antibody [clone: 3B10-5B10]

Lieferant: United States Biological
USBI128966EA 733 CHF
USBI128966
Anti-KRT81 Mouse Monoclonal Antibody [clone: 3B10-5B10]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: KRT81 (keratin 81)
Clonality: monoclonal
Clone: 3B10-5B10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR