Anti-KRT8 Mouse Monoclonal Antibody [clone: 3E3]

Lieferant: United States Biological
USBI128964EA 733 CHF
USBI128964
Anti-KRT8 Mouse Monoclonal Antibody [clone: 3E3]
Antikörper
Type: Primary
Antigen: KRT8 (keratin 8)
Clonality: monoclonal
Clone: 3000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR