Anti-KRT71 Mouse Monoclonal Antibody [clone: 1D4]

Lieferant: United States Biological
USBI128959EA 733 CHF
USBI128959
Anti-KRT71 Mouse Monoclonal Antibody [clone: 1D4]
Antikörper
Type: Primary
Antigen: KRT71 (keratin 71)
Clonality: monoclonal
Clone: 1D4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR