Anti-KRT5 Mouse Monoclonal Antibody [clone: 1A12]

Supplier: US Biological
USBI128958EA 702 CHF
USBI128958
Anti-KRT5 Mouse Monoclonal Antibody [clone: 1A12]
Antibodies
Type: Primary
Antigen: KRT5 (keratin 5)
Clonality: monoclonal
Clone: 1A12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR