Anti-KRT4 Mouse Monoclonal Antibody [clone: 5H5]

Supplier: US Biological
USBI128956EA 702 CHF
USBI128956
Anti-KRT4 Mouse Monoclonal Antibody [clone: 5H5]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: KRT4 (keratin 4)
Clonality: monoclonal
Clone: 5H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR