Anti-KRT34 Mouse Monoclonal Antibody [clone: 3E5]
Lieferant: United States Biological
USBI128951EA 733 CHF
USBI128951
Anti-KRT34 Mouse Monoclonal Antibody [clone: 3E5]
Antikörper
Type: Primary
Antigen: KRT34 (keratin 34)
Clonality: monoclonal
Clone: 300000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human