Anti-KRT23 Mouse Monoclonal Antibody [clone: 2B4-3B5]

Supplier: US Biological
USBI128945EA 702 CHF
USBI128945
Anti-KRT23 Mouse Monoclonal Antibody [clone: 2B4-3B5]
Antibodies
Recommended dilution: Immunochemistry: 5 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KRT23 (keratin 23 (histone deacetylase inducible))
Clonality: monoclonal
Clone: 2B4-3B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR