Anti-KRT13 Mouse Monoclonal Antibody [clone: 4F5]

Supplier: US Biological
USBI128931EA 702 CHF
USBI128931
Anti-KRT13 Mouse Monoclonal Antibody [clone: 4F5]
Antibodies
Type: Primary
Antigen: KRT13 (keratin 13)
Clonality: monoclonal
Clone: 4F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR