Anti-KRT10 Mouse Monoclonal Antibody [clone: 1H6]

Lieferant: United States Biological
USBI128929EA 733 CHF
USBI128929
Anti-KRT10 Mouse Monoclonal Antibody [clone: 1H6]
Antikörper
Type: Primary
Antigen: KRT10 (keratin 10)
Clonality: monoclonal
Clone: 1H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR