Anti-KRT1 Mouse Monoclonal Antibody [clone: 2A7]

Lieferant: United States Biological
USBI128928EA 733 CHF
USBI128928
Anti-KRT1 Mouse Monoclonal Antibody [clone: 2A7]
Antikörper
Type: Primary
Antigen: KRT1 (keratin 1)
Clonality: monoclonal
Clone: 2A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR