Anti-KRR1 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI128927EA 733 CHF
USBI128927
Anti-KRR1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KRR1 (KRR1, small subunit (SSU) processome component, homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR