Anti-KRIT1 Mouse Monoclonal Antibody [clone: 2C7]

Supplier: US Biological
USBI128926EA 702 CHF
USBI128926
Anti-KRIT1 Mouse Monoclonal Antibody [clone: 2C7]
Antibodies
Type: Primary
Antigen: KRIT1 (KRIT1, ankyrin repeat containing)
Clonality: monoclonal
Clone: 2C7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR