Anti-KREMEN1 Mouse Monoclonal Antibody [clone: 1E7]

Supplier: US Biological
USBI128924EA 702 CHF
USBI128924
Anti-KREMEN1 Mouse Monoclonal Antibody [clone: 1E7]
Antibodies
Type: Primary
Antigen: KREMEN1 (kringle containing transmembrane protein 1)
Clonality: monoclonal
Clone: 10000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR