Anti-KRAS Mouse Monoclonal Antibody [clone: 3B10-2F2]

Lieferant: United States Biological
USBI128923EA 733 CHF
USBI128923
Anti-KRAS Mouse Monoclonal Antibody [clone: 3B10-2F2]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KRAS (Kirsten rat sarcoma viral oncogene homolog)
Clonality: monoclonal
Clone: 3B10-2F2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR