Anti-KPNB3 Mouse Monoclonal Antibody [clone: 1C4]

Supplier: US Biological
USBI128920EA 702 CHF
USBI128920
Anti-KPNB3 Mouse Monoclonal Antibody [clone: 1C4]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: KPNB3
Clonality: monoclonal
Clone: 1C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR