Anti-KPNA5 Mouse Monoclonal Antibody [clone: 1D2]

Supplier: US Biological
USBI128918EA 702 CHF
USBI128918
Anti-KPNA5 Mouse Monoclonal Antibody [clone: 1D2]
Antibodies
Type: Primary
Antigen: KPNA5 (karyopherin alpha 5 (importin alpha 6))
Clonality: monoclonal
Clone: 1D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR