Anti-KPNA1 Mouse Monoclonal Antibody [clone: 2A4-1B5]

Supplier: US Biological
USBI128914EA 702 CHF
USBI128914
Anti-KPNA1 Mouse Monoclonal Antibody [clone: 2A4-1B5]
Antibodies
Recommended dilution: Immunochemistry: 1 ?g/ml

Type: Primary
Antigen: KPNA1 (karyopherin alpha 1 (importin alpha 5))
Clonality: monoclonal
Clone: 2A4-1B5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR