Anti-KPNA1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI128913EA 788 CHF
USBI128913
Anti-KPNA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KPNA1 (karyopherin alpha 1 (importin alpha 5))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR