Anti-KNTC1 Mouse Monoclonal Antibody [clone: 10H4]

Lieferant: United States Biological
USBI128911EA 733 CHF
USBI128911
Anti-KNTC1 Mouse Monoclonal Antibody [clone: 10H4]
Antikörper
Recommended dilution: Immunofluorescence: 50 ?g/ml

Type: Primary
Antigen: KNTC1 (kinetochore associated 1)
Clonality: monoclonal
Clone: 10H4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR