Anti-KNG1 Mouse Monoclonal Antibody [clone: 4A1]
Lieferant: United States Biological
USBI128910EA 733 CHF
USBI128910
Anti-KNG1 Mouse Monoclonal Antibody [clone: 4A1]
Antikörper
Type: Primary
Antigen: KNG1 (kininogen 1)
Clonality: monoclonal
Clone: 4A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human