Anti-KLRC4 Mouse Monoclonal Antibody [clone: 1D10]

Supplier: US Biological
USBI128902EA 702 CHF
USBI128902
Anti-KLRC4 Mouse Monoclonal Antibody [clone: 1D10]
Antibodies
Type: Primary
Antigen: KLRC4 (killer cell lectin-like receptor subfamily C, member 4)
Clonality: monoclonal
Clone: 1D10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR