Anti-KLRC3 Mouse Monoclonal Antibody [clone: 3D5]

Supplier: US Biological
USBI128901EA 702 CHF
USBI128901
Anti-KLRC3 Mouse Monoclonal Antibody [clone: 3D5]
Antibodies
Type: Primary
Antigen: KLRC3 (killer cell lectin-like receptor subfamily C, member 3)
Clonality: monoclonal
Clone: 3D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR