Anti-KLRC1 Mouse Monoclonal Antibody [clone: 2C3]

Supplier: US Biological
USBI128899EA 702 CHF
USBI128899
Anti-KLRC1 Mouse Monoclonal Antibody [clone: 2C3]
Antibodies
Type: Primary
Antigen: KLRC1 (killer cell lectin-like receptor subfamily C, member 1)
Clonality: monoclonal
Clone: 2C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR