Anti-KLK8 Mouse Monoclonal Antibody [clone: 2F11]
Lieferant: United States Biological
USBI128894EA 733 CHF
USBI128894
Anti-KLK8 Mouse Monoclonal Antibody [clone: 2F11]
Antikörper
Recommended dilution: Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: KLK8 (kallikrein-related peptidase 8)
Clonality: monoclonal
Clone: 2F11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human