Anti-KLK8 Rabbit Polyclonal Antibody

Supplier: US Biological
USBI128893EA 750 CHF
USBI128893
Anti-KLK8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KLK8 (kallikrein-related peptidase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR