Anti-KLK6 Mouse Monoclonal Antibody [clone: 4A10]

Supplier: US Biological
USBI128888EA 702 CHF
USBI128888
Anti-KLK6 Mouse Monoclonal Antibody [clone: 4A10]
Antibodies
Type: Primary
Antigen: KLK6 (kallikrein-related peptidase 6)
Clonality: monoclonal
Clone: 4A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR