Anti-KLK3 Mouse Monoclonal Antibody [clone: 1B1]

Lieferant: United States Biological
USBI128887EA 733 CHF
USBI128887
Anti-KLK3 Mouse Monoclonal Antibody [clone: 1B1]
Antikörper
Type: Primary
Antigen: KLK3 (kallikrein-related peptidase 3)
Clonality: monoclonal
Clone: 1B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR