Anti-KLK1 Mouse Monoclonal Antibody [clone: 3G2]

Lieferant: United States Biological
USBI128880EA 733 CHF
USBI128880
Anti-KLK1 Mouse Monoclonal Antibody [clone: 3G2]
Antikörper
Type: Primary
Antigen: KLK1 (kallikrein 1)
Clonality: monoclonal
Clone: 3G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR