Anti-KLK1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI128879EA 788 CHF
USBI128879
Anti-KLK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLK1 (kallikrein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR