Anti-KLHL4 Mouse Monoclonal Antibody [clone: 4B6]

Supplier: US Biological
USBI128876EA 702 CHF
USBI128876
Anti-KLHL4 Mouse Monoclonal Antibody [clone: 4B6]
Antibodies
Type: Primary
Antigen: KLHL4 (kelch-like family member 4)
Clonality: monoclonal
Clone: 4B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR