Anti-KLHL25 Mouse Monoclonal Antibody [clone: 1E9]
Lieferant: United States Biological
USBI128871EA 733 CHF
USBI128871
Anti-KLHL25 Mouse Monoclonal Antibody [clone: 1E9]
Antikörper
Type: Primary
Antigen: KLHL25 (kelch-like family member 25)
Clonality: monoclonal
Clone: 1000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human