Anti-KLHL20 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI128870EA 733 CHF
USBI128870
Anti-KLHL20 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLHL20 (kelch-like family member 20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR