Anti-KLHL2 Mouse Monoclonal Antibody [clone: 3G3]

Supplier: US Biological
USBI128869EA 702 CHF
USBI128869
Anti-KLHL2 Mouse Monoclonal Antibody [clone: 3G3]
Antibodies
Type: Primary
Antigen: KLHL2 (kelch-like family member 2)
Clonality: monoclonal
Clone: 3G3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR