Anti-KLHL1 Mouse Monoclonal Antibody [clone: 3C2]

Supplier: US Biological
USBI128866EA 702 CHF
USBI128866
Anti-KLHL1 Mouse Monoclonal Antibody [clone: 3C2]
Antibodies
Type: Primary
Antigen: KLHL1 (kelch-like family member 1)
Clonality: monoclonal
Clone: 3C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR