Anti-KLHDC4 Mouse Monoclonal Antibody [clone: 4G11]

Supplier: US Biological
USBI128865EA 702 CHF
USBI128865
Anti-KLHDC4 Mouse Monoclonal Antibody [clone: 4G11]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 1. 5 ?g/ml

Type: Primary
Antigen: KLHDC4 (kelch domain containing 4)
Clonality: monoclonal
Clone: 4G11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Guinea pig, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR