Anti-KLF9 Mouse Monoclonal Antibody [clone: 1H5]

Lieferant: United States Biological
USBI128862EA 733 CHF
USBI128862
Anti-KLF9 Mouse Monoclonal Antibody [clone: 1H5]
Antikörper
Type: Primary
Antigen: KLF9 (Kruppel-like factor 9)
Clonality: monoclonal
Clone: 1H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR