Anti-KLF8 Mouse Monoclonal Antibody [clone: 2E10]

Supplier: US Biological
USBI128861EA 702 CHF
USBI128861
Anti-KLF8 Mouse Monoclonal Antibody [clone: 2E10]
Antibodies
Type: Primary
Antigen: KLF8 (Kruppel-like factor 8)
Clonality: monoclonal
Clone: 20000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR