Anti-KLF7 Mouse Monoclonal Antibody [clone: 3E8-B8]

Supplier: US Biological
USBI128857EA 702 CHF
USBI128857
Anti-KLF7 Mouse Monoclonal Antibody [clone: 3E8-B8]
Antibodies
Type: Primary
Antigen: KLF7 (Kruppel-like factor 7 (ubiquitous))
Clonality: monoclonal
Clone: 3E8-B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR