Anti-KLF6 Mouse Monoclonal Antibody [clone: 1A9]
Supplier: US Biological
USBI128856EA 702 CHF
USBI128856
Anti-KLF6 Mouse Monoclonal Antibody [clone: 1A9]
Antibodies
Type: Primary
Antigen: KLF6 (Kruppel-like factor 6)
Clonality: monoclonal
Clone: 1A9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human