Anti-KLF5 Mouse Monoclonal Antibody [clone: 2G12]

Supplier: US Biological
USBI128855EA 702 CHF
USBI128855
Anti-KLF5 Mouse Monoclonal Antibody [clone: 2G12]
Antibodies
Type: Primary
Antigen: KLF5 (Kruppel-like factor 5 (intestinal))
Clonality: monoclonal
Clone: 2G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR