Anti-KLF2 Mouse Monoclonal Antibody [clone: 1D1]

Lieferant: United States Biological
USBI128854EA 733 CHF
USBI128854
Anti-KLF2 Mouse Monoclonal Antibody [clone: 1D1]
Antikörper
Type: Primary
Antigen: KLF2 (Kruppel-like factor 2)
Clonality: monoclonal
Clone: 1D1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR