Anti-KLF12 Mouse Monoclonal Antibody [clone: 3D2]

Supplier: US Biological
USBI128849EA 702 CHF
USBI128849
Anti-KLF12 Mouse Monoclonal Antibody [clone: 3D2]
Antibodies
Type: Primary
Antigen: KLF12 (Kruppel-like factor 12)
Clonality: monoclonal
Clone: 3D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR