Anti-KLF11 Mouse Monoclonal Antibody [clone: 8F4]

Supplier: US Biological
USBI128848EA 702 CHF
USBI128848
Anti-KLF11 Mouse Monoclonal Antibody [clone: 8F4]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml; Immunochemistry: 3?g/ml

Type: Primary
Antigen: KLF11 (Kruppel-like factor 11)
Clonality: monoclonal
Clone: 8F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR