Anti-KLF10 Mouse Monoclonal Antibody [clone: 4G9]

Supplier: US Biological
USBI128847EA 702 CHF
USBI128847
Anti-KLF10 Mouse Monoclonal Antibody [clone: 4G9]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: KLF10 (Kruppel-like factor 10)
Clonality: monoclonal
Clone: 4G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR