Anti-KLC1 Mouse Polyclonal Antibody

Lieferant: United States Biological
USBI128843EA 733 CHF
USBI128843
Anti-KLC1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLC1 (kinesin light chain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR