Anti-KISS1 Mouse Monoclonal Antibody [clone: 1F7]

Supplier: US Biological
USBI128842EA 702 CHF
USBI128842
Anti-KISS1 Mouse Monoclonal Antibody [clone: 1F7]
Antibodies
Recommended dilution: Immunochemistry: 3?g/ml

Type: Primary
Antigen: KISS1 (KiSS-1 metastasis-suppressor)
Clonality: monoclonal
Clone: 1F7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR