Anti-KIRREL3 Mouse Monoclonal Antibody [clone: 3A12]

Supplier: US Biological
USBI128840EA 702 CHF
USBI128840
Anti-KIRREL3 Mouse Monoclonal Antibody [clone: 3A12]
Antibodies
Type: Primary
Antigen: KIRREL3 (kin of IRRE like 3 (Drosophila))
Clonality: monoclonal
Clone: 3A12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR