Anti-KIR3DX1 Mouse Monoclonal Antibody [clone: 3E7]

Supplier: US Biological
USBI128837EA 702 CHF
USBI128837
Anti-KIR3DX1 Mouse Monoclonal Antibody [clone: 3E7]
Antibodies
Type: Primary
Antigen: KIR3DX1 (killer cell immunoglobulin-like receptor, three domains, X1)
Clonality: monoclonal
Clone: 30000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR